Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135774.1 | 7585 | Intron | NP_001129246.1 | ||
NM_003490.3 | 7585 | Intron | NP_003481.3 | ||
NM_133633.2 | 7585 | Intron | NP_598344.2 | ||
XM_011530405.2 | 7585 | UTR 3 | XP_011528707.1 | ||
XM_011530406.2 | 7585 | UTR 3 | XP_011528708.1 | ||
XM_011530407.2 | 7585 | UTR 3 | XP_011528709.1 | ||
XM_011530408.1 | 7585 | UTR 3 | XP_011528710.1 | ||
XM_011530410.2 | 7585 | UTR 3 | XP_011528712.1 | ||
XM_017028960.1 | 7585 | UTR 3 | XP_016884449.1 | ||
XM_017028961.1 | 7585 | UTR 3 | XP_016884450.1 | ||
XM_017028962.1 | 7585 | UTR 3 | XP_016884451.1 | ||
XM_017028963.1 | 7585 | UTR 3 | XP_016884452.1 | ||
XM_017028964.1 | 7585 | UTR 3 | XP_016884453.1 | ||
XM_017028965.1 | 7585 | UTR 3 | XP_016884454.1 |