Product Details

SNP ID
rs8140217
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38821996 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGGTGGGGCTGGGCCAGGCATG[A/G]AACCAACATTCAAACCGCTACACAC
Phenotype
MIM: 609474
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NPTXR PubMed Links
Additional Information
For this assay, SNP(s) [rs138908300] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NPTXR
Gene Name
neuronal pentraxin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014293.3 2270 UTR 3 NP_055108.2

View Full Product Details