Product Details

SNP ID
rs11198227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118007183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATGTTTAAAAAAAAAAAAAAAA[C/G]TACCAAGACCTCAAACGGGAAAATA
Phenotype
MIM: 608599
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB11FIP2 PubMed Links

Gene Details

Gene
RAB11FIP2
Gene Name
RAB11 family interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014904.2 3794 UTR 3 NP_055719.1
XM_005269629.3 3794 UTR 3 XP_005269686.1
XM_017015926.1 3794 Intron XP_016871415.1

View Full Product Details