Product Details

SNP ID
rs6584499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102428502 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGGGCATTTTTCTAGGTGCTAGG[G/T]TACACAAACTATTTCCTCTGCCTGG
Phenotype
MIM: 614142 MIM: 610287 MIM: 610567 MIM: 602327
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CUEDC2 PubMed Links

Gene Details

Gene
CUEDC2
Gene Name
CUE domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024040.2 Intron NP_076945.2
Gene
FBXL15
Gene Name
F-box and leucine rich repeat protein 15
There are no transcripts associated with this gene.

Gene
MIR146B
Gene Name
microRNA 146b
There are no transcripts associated with this gene.

Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
There are no transcripts associated with this gene.

View Full Product Details