Product Details

SNP ID
rs11190490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100374409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGATGTGTGAGATGAGTGAGG[A/T]CAGCAATCAGAAGAGGACTTTGAGG
Phenotype
MIM: 616240 MIM: 604031
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OLMALINC PubMed Links

Gene Details

Gene
OLMALINC
Gene Name
oligodendrocyte maturation-associated long intergenic non-coding RNA
There are no transcripts associated with this gene.

Gene
SCD
Gene Name
stearoyl-CoA desaturase
There are no transcripts associated with this gene.

View Full Product Details