Product Details

SNP ID
rs9332092
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.10:94936772 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
GTTTCATGAGTCAGGGACCAAGTTA[C/T]TGCTTTTCTTTGCCCTGTATAAAGG
Phenotype
MIM: 601130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
CYP2C9*18,g.-1911T>C CYP2C9*3A,g.-1911T>C CYP2C9*3B,g.-1911T>C
Literature Links
CYP2C9 PubMed Links

Gene Details

Gene
CYP2C9
Gene Name
cytochrome P450 family 2 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000771.3 Intron NP_000762.2
XM_017015758.1 Intron XP_016871247.1

View Full Product Details