Product Details

SNP ID
rs11919985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:72729188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACTTGTGTGCCAGGAGCATAAA[C/T]AGTCACGGTTGCCCCTGGATTCGTT
Phenotype
MIM: 613663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHQ1 PubMed Links

Gene Details

Gene
SHQ1
Gene Name
SHQ1, H/ACA ribonucleoprotein assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018130.2 Intron NP_060600.2
XM_011533895.2 Intron XP_011532197.1
XM_011533896.2 Intron XP_011532198.1
XM_011533897.2 Intron XP_011532199.1
XM_011533898.2 Intron XP_011532200.1
XM_011533899.1 Intron XP_011532201.1
XM_017006724.1 Intron XP_016862213.1

View Full Product Details