Product Details

SNP ID
rs4380112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79071142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGATTAGGTGTTGGTATAGTTTT[C/T]ACAGTGCTTGCTCTGGGTATTACAA
Phenotype
MIM: 614446
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP9B PubMed Links

Gene Details

Gene
ATP9B
Gene Name
ATPase phospholipid transporting 9B (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306085.1 Intron NP_001293014.1
NM_198531.4 Intron NP_940933.3
XM_011525963.2 Intron XP_011524265.1
XM_011525964.2 Intron XP_011524266.1
XM_011525965.2 Intron XP_011524267.1
XM_011525966.2 Intron XP_011524268.1
XM_011525971.2 Intron XP_011524273.2
XM_011525972.2 Intron XP_011524274.1
XM_011525973.2 Intron XP_011524275.1
XM_011525974.2 Intron XP_011524276.1
XM_017025726.1 Intron XP_016881215.1
XM_017025727.1 Intron XP_016881216.1
XM_017025728.1 Intron XP_016881217.1
XM_017025729.1 Intron XP_016881218.1
XM_017025730.1 Intron XP_016881219.1
XM_017025731.1 Intron XP_016881220.1
XM_017025732.1 Intron XP_016881221.1
XM_017025733.1 Intron XP_016881222.1
XM_017025734.1 Intron XP_016881223.1
XM_017025735.1 Intron XP_016881224.1
XM_017025736.1 Intron XP_016881225.1
XM_017025737.1 Intron XP_016881226.1
XM_017025738.1 Intron XP_016881227.1
XM_017025739.1 Intron XP_016881228.1
XM_017025740.1 Intron XP_016881229.1
XM_017025741.1 Intron XP_016881230.1
XM_017025742.1 Intron XP_016881231.1

View Full Product Details