Product Details

SNP ID
rs11213824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111305833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTGTACTTCCGAGCCAATAGTG[C/T]TAATTGGTACTCTATGTTCCTGGTT
Phenotype
MIM: 615693 MIM: 615694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COLCA1 PubMed Links

Gene Details

Gene
COLCA1
Gene Name
colorectal cancer associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302644.1 Intron NP_001289573.1
NM_001302645.1 Intron NP_001289574.1
NM_001302646.1 Intron NP_001289575.1
NM_001302647.1 Intron NP_001289576.1
NM_001302648.1 Intron NP_001289577.1
NM_207429.3 Intron NP_997312.1
Gene
COLCA2
Gene Name
colorectal cancer associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136105.2 Intron NP_001129577.1
NM_001271457.1 Intron NP_001258386.1
NM_001271458.1 Intron NP_001258387.1
XM_017017196.1 Intron XP_016872685.1
XM_017017197.1 Intron XP_016872686.1
XM_017017198.1 Intron XP_016872687.1
XM_017017199.1 Intron XP_016872688.1
XM_017017200.1 Intron XP_016872689.1

View Full Product Details