Product Details
- SNP ID
-
rs12272373
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:35142427 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAATGTCCAACAATGATAGACTGG[A/G]TGAGGGGATCTTTGCTTGGGAATTG
- Phenotype
-
MIM: 107269
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD44
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138781086] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD44
- Gene Name
- CD44 molecule (Indian blood group)
- Gene
- LOC100507144
- Gene Name
- uncharacterized LOC100507144
There are no transcripts associated with this gene.
View Full Product Details