Product Details

SNP ID
rs12362696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5806804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGCAGTAGATTGTTTTGGGAG[A/G]TAGTTTGGACTTACTAGGGTAATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52N2 PubMed Links

Gene Details

Gene
OR52N2
Gene Name
olfactory receptor family 52 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005174.1 3372 Intron NP_001005174.1
XM_011520090.2 3372 UTR 5 XP_011518392.1

View Full Product Details