Product Details

SNP ID
rs11039102
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5806023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAATAAAGCGTATTTTAAAGAGG[C/T]CTACAGGGTCGTATCCATTGGCCTC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR52N2 PubMed Links

Gene Details

Gene
OR52N2
Gene Name
olfactory receptor family 52 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005174.1 2591 Intron NP_001005174.1
XM_011520090.2 2591 UTR 5 XP_011518392.1

View Full Product Details