Product Details

SNP ID
rs10837375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5121040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCACGCGTTCACAGTAAGTGTGG[G/T]ATATTAACTTGGTTCGGTAGAGTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52A4P PubMed Links

Gene Details

Gene
OR52A4P
Gene Name
olfactory receptor family 52 subfamily A member 4 pseudogene
There are no transcripts associated with this gene.

View Full Product Details