Product Details

SNP ID
rs12979224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7678394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGGGCTTTAAAAGGGAGCTT[G/T]GGAATGGTGAGCGGAGGGTCTGAGG
Phenotype
MIM: 609565 MIM: 605565
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MCEMP1 PubMed Links

Gene Details

Gene
MCEMP1
Gene Name
mast cell expressed membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174918.2 392 Missense Mutation GGG,TGG G123W NP_777578.2
Gene
RETN
Gene Name
resistin
There are no transcripts associated with this gene.

Gene
TRAPPC5
Gene Name
trafficking protein particle complex 5
There are no transcripts associated with this gene.

View Full Product Details