Product Details

SNP ID
rs12462152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1417398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTGCACAAGGACGTTTGCGTCA[C/G]CTGCTTCTGTGGTTTGAATTAAGAC
Phenotype
MIM: 607430
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DAZAP1 PubMed Links

Gene Details

Gene
DAZAP1
Gene Name
DAZ associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018959.3 Intron NP_061832.2
NM_170711.2 Intron NP_733829.1
XM_005259531.3 Intron XP_005259588.1
XM_005259534.3 Intron XP_005259591.1
XM_005259535.3 Intron XP_005259592.1
XM_005259536.3 Intron XP_005259593.1
XM_011527904.2 Intron XP_011526206.1
XM_011527905.2 Intron XP_011526207.1
XM_011527906.2 Intron XP_011526208.1
XM_011527907.2 Intron XP_011526209.1
XM_011527908.2 Intron XP_011526210.1
XM_011527909.2 Intron XP_011526211.1
XM_011527910.2 Intron XP_011526212.1
XM_017026581.1 Intron XP_016882070.1
XM_017026582.1 Intron XP_016882071.1

View Full Product Details