Product Details

SNP ID
rs13389370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:229923426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGGCTGAGTTTACGCCTGTCA[A/C]TGAGTTATTCATGCACCCTGCCCTT
Phenotype
MIM: 609105 MIM: 604506
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FBXO36 PubMed Links

Gene Details

Gene
FBXO36
Gene Name
F-box protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174899.4 Intron NP_777559.3
XM_005246317.1 Intron XP_005246374.1
Gene
TRIP12
Gene Name
thyroid hormone receptor interactor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284214.1 Intron NP_001271143.1
NM_001284215.1 Intron NP_001271144.1
NM_001284216.1 Intron NP_001271145.1
NM_004238.2 Intron NP_004229.1
XM_005246954.4 Intron XP_005247011.1
XM_005246955.4 Intron XP_005247012.1
XM_005246956.4 Intron XP_005247013.1
XM_005246957.4 Intron XP_005247014.1
XM_005246958.4 Intron XP_005247015.1
XM_005246960.4 Intron XP_005247017.1
XM_005246961.3 Intron XP_005247018.1
XM_005246962.4 Intron XP_005247019.1
XM_006712852.3 Intron XP_006712915.1
XM_011512180.2 Intron XP_011510482.1
XM_011512181.2 Intron XP_011510483.1
XM_011512183.2 Intron XP_011510485.1
XM_017005272.1 Intron XP_016860761.1
XM_017005273.1 Intron XP_016860762.1
XM_017005274.1 Intron XP_016860763.1
XM_017005275.1 Intron XP_016860764.1
XM_017005276.1 Intron XP_016860765.1
XM_017005277.1 Intron XP_016860766.1
XM_017005278.1 Intron XP_016860767.1
XM_017005279.1 Intron XP_016860768.1
XM_017005280.1 Intron XP_016860769.1
XM_017005281.1 Intron XP_016860770.1
XM_017005282.1 Intron XP_016860771.1
XM_017005283.1 Intron XP_016860772.1
XM_017005284.1 Intron XP_016860773.1
XM_017005285.1 Intron XP_016860774.1
XM_017005286.1 Intron XP_016860775.1
XM_017005287.1 Intron XP_016860776.1
XM_017005288.1 Intron XP_016860777.1
XM_017005289.1 Intron XP_016860778.1
XM_017005290.1 Intron XP_016860779.1
XM_017005291.1 Intron XP_016860780.1
XM_017005292.1 Intron XP_016860781.1
XM_017005293.1 Intron XP_016860782.1

View Full Product Details