Product Details
- SNP ID
-
rs9333321
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:43155585 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGCTAGTGGCCCCACACTGATACC[C/T]ACAACCCCTCTCTTCTCTGGAGTTT
- Phenotype
-
MIM: 109610
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPO
PubMed Links
Gene Details
- Gene
- TSPO
- Gene Name
- translocator protein
View Full Product Details