Product Details

SNP ID
rs11231747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64319488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCGACACATGGGGAAACCTTC[C/T]CTCCCTTTCTCTGTTGTCACCAATG
Phenotype
MIM: 601998 MIM: 606583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRRA PubMed Links

Gene Details

Gene
ESRRA
Gene Name
estrogen related receptor alpha
There are no transcripts associated with this gene.

Gene
PRDX5
Gene Name
peroxiredoxin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012094.4 Intron NP_036226.1
NM_181651.2 Intron NP_857634.1
NM_181652.2 Intron NP_857635.1
XM_005273882.4 Intron XP_005273939.2
Gene
TRMT112
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details