Product Details

SNP ID
rs6952735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100365950 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTAGGATGGTAAATTCTAGGTGG[G/T]TTTTTTTTTTTTTTTTTTTTTTAGA
Phenotype
MIM: 605341 MIM: 605342
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR6840 PubMed Links

Gene Details

Gene
MIR6840
Gene Name
microRNA 6840
There are no transcripts associated with this gene.

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
There are no transcripts associated with this gene.

Gene
PILRB
Gene Name
paired immunoglobin-like type 2 receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178238.3 Intron NP_839956.1
Gene
STAG3L5P-PVRIG2P-PILRB
Gene Name
STAG3L5P-PVRIG2P-PILRB readthrough
There are no transcripts associated with this gene.

View Full Product Details