Product Details

SNP ID
rs12386703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92528488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCCTAGCGCCGCAAAGGACCCG[C/G]GACCCGGCAGGCCGAGGACGTCGGA
Phenotype
MIM: 602136
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PEX1 PubMed Links

Gene Details

Gene
PEX1
Gene Name
peroxisomal biogenesis factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000466.2 44 UTR 5 NP_000457.1
NM_001282677.1 44 UTR 5 NP_001269606.1
NM_001282678.1 44 UTR 5 NP_001269607.1
XM_017012319.1 44 Intron XP_016867808.1
Gene
RBM48
Gene Name
RNA binding motif protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032120.3 44 Intron NP_115496.2
XM_005250636.4 44 Intron XP_005250693.1
XM_006716149.3 44 Intron XP_006716212.1
XM_006716150.3 44 Intron XP_006716213.1
XM_017012709.1 44 Intron XP_016868198.1

View Full Product Details