Product Details

SNP ID
rs12055925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92200068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCATGTTGACGTTCAGTATATT[G/T]TGAAATACACATGAAGTTATACCAA
Phenotype
MIM: 604214
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRIT1 PubMed Links

Gene Details

Gene
KRIT1
Gene Name
KRIT1, ankyrin repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013406.1 3618 UTR 3 NP_001013424.1
NM_004912.3 3618 UTR 3 NP_004903.2
NM_194454.1 3618 UTR 3 NP_919436.1
NM_194455.1 3618 UTR 3 NP_919437.1
NM_194456.1 3618 UTR 3 NP_919438.1
XM_005250660.3 3618 UTR 3 XP_005250717.1
XM_005250662.3 3618 UTR 3 XP_005250719.1
XM_005250665.3 3618 UTR 3 XP_005250722.1
XM_005250666.3 3618 UTR 3 XP_005250723.1
XM_005250667.2 3618 UTR 3 XP_005250724.1
XM_005250668.3 3618 UTR 3 XP_005250725.1
XM_005250669.3 3618 UTR 3 XP_005250726.1
XM_006716161.3 3618 UTR 3 XP_006716224.1
XM_006716162.3 3618 UTR 3 XP_006716225.1
XM_006716163.3 3618 UTR 3 XP_006716226.1
XM_011516651.2 3618 UTR 3 XP_011514953.1
XM_011516653.2 3618 UTR 3 XP_011514955.1
XM_011516654.2 3618 UTR 3 XP_011514956.1
XM_011516655.2 3618 UTR 3 XP_011514957.1
XM_011516656.2 3618 UTR 3 XP_011514958.1
XM_011516657.2 3618 UTR 3 XP_011514959.1
XM_011516658.2 3618 UTR 3 XP_011514960.1
XM_011516659.2 3618 UTR 3 XP_011514961.1
XM_011516660.2 3618 UTR 3 XP_011514962.1
XM_011516661.2 3618 UTR 3 XP_011514963.1
XM_017012755.1 3618 UTR 3 XP_016868244.1
XM_017012756.1 3618 UTR 3 XP_016868245.1
XM_017012757.1 3618 UTR 3 XP_016868246.1
XM_017012758.1 3618 UTR 3 XP_016868247.1
XM_017012759.1 3618 UTR 3 XP_016868248.1
Gene
MIR1285-1
Gene Name
microRNA 1285-1
There are no transcripts associated with this gene.

View Full Product Details