Product Details
- SNP ID
-
rs12893595
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:95183066 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTGAGATAAATAAAAACTACCTTC[A/G]TAACCTTTGGATTTTTAAACTTAGA
- Phenotype
-
MIM: 611121
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLMN
PubMed Links
Gene Details
- Gene
- CLMN
- Gene Name
- calmin
- Gene
- DICER1-AS1
- Gene Name
- DICER1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details