Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142308.2 | Intron | NP_001135780.2 | |||
XM_011519453.2 | Intron | XP_011517755.1 | |||
XM_011519454.1 | Intron | XP_011517756.1 | |||
XM_011519455.2 | Intron | XP_011517757.1 | |||
XM_017016182.1 | Intron | XP_016871671.1 | |||
XM_017016183.1 | Intron | XP_016871672.1 | |||
XM_017016184.1 | Intron | XP_016871673.1 | |||
XM_017016185.1 | Intron | XP_016871674.1 | |||
XM_017016186.1 | Intron | XP_016871675.1 |