Product Details

SNP ID
rs12355433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7757717 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTAAAGGAATTAAGTTCCCAGAT[A/G]AAGTATAACCTAGGTTAGATTTTTC
Phenotype
MIM: 146640 MIM: 601720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH2 PubMed Links

Gene Details

Gene
ITIH2
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
There are no transcripts associated with this gene.

Gene
KIN
Gene Name
Kin17 DNA and RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012311.3 Intron NP_036443.1
XM_006717434.3 Intron XP_006717497.1
XM_006717435.3 Intron XP_006717498.1
XM_011519427.1 Intron XP_011517729.1
XM_017015946.1 Intron XP_016871435.1

View Full Product Details