Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134395.1 | Intron | NP_001127867.1 | |||
NM_001134396.1 | Intron | NP_001127868.1 | |||
NM_001318252.1 | Intron | NP_001305181.1 | |||
NM_032350.5 | Intron | NP_115726.1 | |||
XM_006715793.2 | Intron | XP_006715856.1 | |||
XM_011515580.2 | Intron | XP_011513882.1 | |||
XM_011515581.2 | Intron | XP_011513883.1 | |||
XM_011515582.2 | Intron | XP_011513884.1 | |||
XM_011515583.2 | Intron | XP_011513885.1 | |||
XM_011515584.2 | Intron | XP_011513886.1 | |||
XM_017012719.1 | Intron | XP_016868208.1 | |||
XM_017012720.1 | Intron | XP_016868209.1 | |||
XM_017012721.1 | Intron | XP_016868210.1 | |||
XM_017012722.1 | Intron | XP_016868211.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_017781.2 | Intron | NP_060251.2 | |||
XM_011515440.2 | Intron | XP_011513742.1 | |||
XM_011515441.2 | Intron | XP_011513743.1 |