Product Details

SNP ID
rs10985685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122548381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATAAAAATGATAGTTTCCTCTA[A/G]TACAGGGAGGAAATTTTCCACAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1L8 PubMed Links

Gene Details

Gene
OR1L8
Gene Name
olfactory receptor family 1 subfamily L member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004454.1 Intron NP_001004454.1
XM_017014284.1 Intron XP_016869773.1
XM_017014285.1 Intron XP_016869774.1
XM_017014286.1 Intron XP_016869775.1
Gene
OR1N2
Gene Name
olfactory receptor family 1 subfamily N member 2
There are no transcripts associated with this gene.

View Full Product Details