Product Details

SNP ID
rs12345709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121816333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGTGTTCTGTGGCCCAAGACTG[A/G]GTCCTGGTTGGAATCTTAGCAAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107987125 PubMed Links

Gene Details

Gene
LOC107987125
Gene Name
uncharacterized LOC107987125
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017015410.1 8146 Intron XP_016870899.1
Gene
TTLL11
Gene Name
tubulin tyrosine ligase like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139442.1 8146 Intron NP_001132914.1
NM_194252.2 8146 Intron NP_919228.2
XM_005251728.2 8146 UTR 3 XP_005251785.1
XM_005251729.4 8146 UTR 3 XP_005251786.1
XM_006716966.3 8146 Intron XP_006717029.1
XM_017014309.1 8146 Intron XP_016869798.1

View Full Product Details