Product Details

SNP ID
rs11541836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129084678 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTGTCTTGCCAGGTGATCTCTC[C/T]GGCCACCTCCTTGCCTACCTGAGCC
Phenotype
MIM: 614516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOLPP1 PubMed Links

Gene Details

Gene
DOLPP1
Gene Name
dolichyldiphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135917.1 115 Silent Mutation TCC,TCT S29S NP_001129389.1
NM_020438.4 115 Silent Mutation TCC,TCT S29S NP_065171.2

View Full Product Details