Product Details

SNP ID
rs10988468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129672425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACTGAGGTGTGAGGTGAAGGTC[A/C]CACCGTTCCCATTCCTGTAAGATTC
Phenotype
MIM: 604675
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRRX2 PubMed Links

Gene Details

Gene
PRRX2
Gene Name
paired related homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016307.3 Intron NP_057391.1
XM_017014803.1 Intron XP_016870292.1

View Full Product Details