Product Details

SNP ID
rs10849440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6313106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGGAGCTGTGTGGCTGTGGC[C/T]TCAGCCTTTCACACAGAAGGGCAGC
Phenotype
MIM: 611743
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG6 PubMed Links

Gene Details

Gene
PLEKHG6
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144856.1 456 Intron NP_001138328.1
NM_001144857.1 456 UTR 5 NP_001138329.1
NM_018173.3 456 Intron NP_060643.2
XM_005253704.4 456 Intron XP_005253761.1
XM_006718985.3 456 Intron XP_006719048.1
XM_011520967.2 456 Intron XP_011519269.1
XM_017019555.1 456 Intron XP_016875044.1

View Full Product Details