Product Details

SNP ID
rs12315570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2827120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGCAGACACCATAGTCCCCA[C/G]CTACCTGGCTTCAAGAGCCCCCGTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITFG2 PubMed Links

Gene Details

Gene
ITFG2
Gene Name
integrin alpha FG-GAP repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018463.3 947 Intron NP_060933.3
XM_005253707.4 947 Intron XP_005253764.1
XM_005253709.4 947 Intron XP_005253766.1
XM_017019641.1 947 Intron XP_016875130.1
Gene
LOC100507424
Gene Name
uncharacterized LOC100507424
There are no transcripts associated with this gene.

Gene
NRIP2
Gene Name
nuclear receptor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031474.2 947 UTR 3 NP_113662.1
XM_005253797.3 947 Intron XP_005253854.1
XM_005253798.3 947 Intron XP_005253855.1
XM_011521031.2 947 UTR 3 XP_011519333.1
XM_017020000.1 947 UTR 3 XP_016875489.1
XM_017020001.1 947 Intron XP_016875490.1

View Full Product Details