Product Details

SNP ID
rs9333372
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:165631529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGGGAGCGTCTAAGCGAGGCCCG[A/G]CCTATTTATGTGTAACTCAGGAAAA
Phenotype
MIM: 604564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGST3 PubMed Links

Gene Details

Gene
MGST3
Gene Name
microsomal glutathione S-transferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004528.3 Intron NP_004519.1
XM_005245174.3 Intron XP_005245231.2

View Full Product Details