Product Details

SNP ID
rs12069064
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:167726457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTAGTGCTTAACACAGTGGATATG[C/G]TAGATGCTAAAAATATTTGTTAAAT
Phenotype
MIM: 604376
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MPZL1 PubMed Links

Gene Details

Gene
MPZL1
Gene Name
myelin protein zero like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146191.1 Intron NP_001139663.1
NM_003953.5 Intron NP_003944.1
NM_024569.4 Intron NP_078845.3

View Full Product Details