Product Details
- SNP ID
-
rs12121361
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:168182197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACTAAGGTGCTAAATAAATTGGT[G/T]GGGTAAAATATTGGATTAAAGAAGC
- Phenotype
-
MIM: 611807
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TIPRL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75562105] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TIPRL
- Gene Name
- TOR signaling pathway regulator
View Full Product Details