Product Details
- SNP ID
-
rs12134975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:196656867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTATCTTGTTTTATCTCTAATTAT[A/G]ACATACTTGTTATTCTATCCAACTT
- Phenotype
-
MIM: 134370
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFH
PubMed Links
Gene Details
- Gene
- CFH
- Gene Name
- complement factor H
View Full Product Details