Product Details
- SNP ID
-
rs13253402
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:101054497 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTTATTTACTTCTCTGTCTCCCA[C/G]ATAACCCATGGGCTCCTGAGAGCAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ42969
PubMed Links
Gene Details
- Gene
- FLJ42969
- Gene Name
- uncharacterized LOC441374
There are no transcripts associated with this gene.
View Full Product Details