Product Details

SNP ID
rs13437725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27107283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAACAAAAGGCCCAAGAACCCTT[C/T]TGAACCATCAGGGTACAGAATTCTT
Phenotype
MIM: 604685 MIM: 142954
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOTAIRM1 PubMed Links

Gene Details

Gene
HOTAIRM1
Gene Name
HOXA transcript antisense RNA, myeloid-specific 1
There are no transcripts associated with this gene.

Gene
HOXA2
Gene Name
homeobox A2
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030661.4 2867 UTR 3 NP_109377.1
NM_153631.2 2867 UTR 3 NP_705895.1
XM_005249730.2 2867 UTR 3 XP_005249787.1
XM_005249731.2 2867 UTR 3 XP_005249788.1
XM_005249732.3 2867 UTR 3 XP_005249789.1
XM_006715715.2 2867 UTR 3 XP_006715778.1
XM_011515343.2 2867 UTR 3 XP_011513645.1

View Full Product Details