Product Details
- SNP ID
-
rs12996213
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:121220881 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCATAAGCTATAAGCTTTAACT[A/C]CAACCACGAGAAGGTAAGAAGAGCA
- Phenotype
-
MIM: 609785
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TFCP2L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116740749] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFCP2L1
- Gene Name
- transcription factor CP2-like 1
View Full Product Details