Product Details

SNP ID
rs13394108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127702902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTCTTCGGAAATCCTGCAAATAG[A/G]AAGATAATTCTAGATCCGGAATACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFT2D3 PubMed Links

Gene Details

Gene
SFT2D3
Gene Name
SFT2 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032740.3 1880 UTR 3 NP_116129.3
Gene
WDR33
Gene Name
WD repeat domain 33
There are no transcripts associated with this gene.

View Full Product Details