Product Details

SNP ID
rs11514185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56738858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCACCGCGCCTCACCACGAACAA[C/G]AGAAGTTTAATGTTGGTTTCTGATC
Phenotype
MIM: 176635
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRIM1 PubMed Links

Gene Details

Gene
PRIM1
Gene Name
primase (DNA) subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000946.2 Intron NP_000937.1

View Full Product Details