Product Details

SNP ID
rs12861895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85256270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATCATAAACTCAGATAATGTCC[C/T]ACAGTTCTCCCTGTGTTTATATTGC
Phenotype
MIM: 314990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF711 PubMed Links

Gene Details

Gene
ZNF711
Gene Name
zinc finger protein 711
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021998.4 Intron NP_068838.3
XM_005262186.2 Intron XP_005262243.1
XM_005262187.1 Intron XP_005262244.1
XM_005262188.1 Intron XP_005262245.1
XM_005262189.2 Intron XP_005262246.1
XM_011531019.1 Intron XP_011529321.1
XM_011531020.1 Intron XP_011529322.1
XM_011531021.1 Intron XP_011529323.1
XM_011531022.1 Intron XP_011529324.1
XM_011531023.2 Intron XP_011529325.1
XM_011531024.1 Intron XP_011529326.1
XM_011531025.1 Intron XP_011529327.1
XM_011531026.1 Intron XP_011529328.1
XM_017029803.1 Intron XP_016885292.1
XM_017029804.1 Intron XP_016885293.1
XM_017029805.1 Intron XP_016885294.1
XM_017029806.1 Intron XP_016885295.1
XM_017029807.1 Intron XP_016885296.1
XM_017029808.1 Intron XP_016885297.1
XM_017029809.1 Intron XP_016885298.1

View Full Product Details