Product Details

SNP ID
rs11798017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:6770582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGTCAGCACACATCACACAGC[C/T]GCACATTCTCTAATGCAAACTAACC
Phenotype
MIM: 306480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PUDP PubMed Links

Gene Details

Gene
PUDP
Gene Name
pseudouridine 5'-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135565.1 Intron NP_001129037.1
NM_001178135.1 Intron NP_001171606.1
NM_001178136.1 Intron NP_001171607.1
NM_012080.4 Intron NP_036212.3

View Full Product Details