Product Details

SNP ID
rs12917072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80155467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAAGCATCATGCTCCAGATCTC[A/C]CAGCCAGTAAGAGGCAGATATGGGG
Phenotype
MIM: 613871
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAH PubMed Links

Gene Details

Gene
FAH
Gene Name
fumarylacetoacetate hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000137.2 Intron NP_000128.1

View Full Product Details