Product Details
- SNP ID
-
rs6901782
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:42991162 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTTCTTTCCTACCTCTGTGTACT[C/T]GTGCAGTTGGTTCATAGTAAATGCT
- Phenotype
-
MIM: 143170
MIM: 601646
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MEA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199508198] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MEA1
- Gene Name
- male-enhanced antigen 1
- Gene
- PPP2R5D
- Gene Name
- protein phosphatase 2 regulatory subunit B'delta
View Full Product Details