Product Details

SNP ID
rs9333510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43583199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGTCGAAAATAATACTCCATGA[A/G]GCTAGGAACTGGTGTTTTGAATTTG
Phenotype
MIM: 603968 MIM: 607845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLH PubMed Links

Gene Details

Gene
POLH
Gene Name
DNA polymerase eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291969.1 Intron NP_001278898.1
NM_001291970.1 Intron NP_001278899.1
NM_006502.2 Intron NP_006493.1
XM_005249186.3 Intron XP_005249243.1
XM_011514698.2 Intron XP_011513000.1
Gene
XPO5
Gene Name
exportin 5
There are no transcripts associated with this gene.

View Full Product Details