Product Details

SNP ID
rs12431132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:21679608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATAAAATGTAGCACTTTAATTT[A/C]CAATAAAAAGGAAAAAATGGCTGAG
Phenotype
MIM: 600921
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGF9 PubMed Links

Gene Details

Gene
FGF9
Gene Name
fibroblast growth factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002010.2 Intron NP_002001.1
XM_011534996.2 Intron XP_011533298.1

View Full Product Details