Product Details
- SNP ID
-
rs12665231
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:155395463 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTAAGGAAGTATGGTATAGTGTAC[C/T]GTTCACAATAGTTAATTATTATTCA
- Phenotype
-
MIM: 607105
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOX3
PubMed Links
Gene Details
- Gene
- NOX3
- Gene Name
- NADPH oxidase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015718.2 |
1949 |
UTR 3 |
|
|
NP_056533.1 |
View Full Product Details