Product Details
- SNP ID
-
rs9557352
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:99983529 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTTGTCTGAATGTGCTTTTCTG[C/T]TCGGAGTGTATGTCTGTCTGAGTGG
- Phenotype
-
MIM: 603073
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZIC2
PubMed Links
Gene Details
- Gene
- ZIC2
- Gene Name
- Zic family member 2
View Full Product Details