Product Details
- SNP ID
-
rs13049404
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46311485 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTAATACTTCCTGACCGTTGAGTA[A/G]GGAAGGCCTGGGGAGGAGGAATCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C21orf58
PubMed Links
Gene Details
- Gene
- C21orf58
- Gene Name
- chromosome 21 open reading frame 58
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286462.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
NP_001273391.1 |
NM_001286463.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
NP_001273392.1 |
NM_001286476.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
NP_001273405.1 |
NM_001286477.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
NP_001273406.1 |
NM_058180.4 |
2103 |
Missense Mutation |
CCT,CTT |
P231L |
NP_478060.2 |
XM_005261148.4 |
2103 |
Missense Mutation |
CCT,CTT |
P231L |
XP_005261205.1 |
XM_006724018.3 |
2103 |
Missense Mutation |
CCT,CTT |
P231L |
XP_006724081.1 |
XM_011529619.2 |
2103 |
Missense Mutation |
CCT,CTT |
P231L |
XP_011527921.1 |
XM_011529620.2 |
2103 |
Missense Mutation |
CCT,CTT |
P231L |
XP_011527922.1 |
XM_011529621.2 |
2103 |
Missense Mutation |
CCT,CTT |
P230L |
XP_011527923.1 |
XM_011529623.2 |
2103 |
Missense Mutation |
CCT,CTT |
P204L |
XP_011527925.1 |
XM_011529624.2 |
2103 |
Missense Mutation |
CCT,CTT |
P166L |
XP_011527926.1 |
XM_011529625.2 |
2103 |
Missense Mutation |
CCT,CTT |
P148L |
XP_011527927.1 |
XM_011529626.2 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_011527928.1 |
XM_011529627.2 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_011527929.1 |
XM_011529628.1 |
2103 |
Intron |
|
|
XP_011527930.1 |
XM_017028388.1 |
2103 |
Missense Mutation |
CCT,CTT |
P148L |
XP_016883877.1 |
XM_017028389.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_016883878.1 |
XM_017028390.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_016883879.1 |
XM_017028391.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_016883880.1 |
XM_017028392.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_016883881.1 |
XM_017028393.1 |
2103 |
Missense Mutation |
CCT,CTT |
P125L |
XP_016883882.1 |
- Gene
- YBEY
- Gene Name
- ybeY metallopeptidase (putative)
View Full Product Details