Product Details
- SNP ID
-
rs13003319
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:176145798 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGGAGCCAAGTGTTAATTCCAA[C/T]TGCTACTCCCACCCCCAAACCCACC
- Phenotype
-
MIM: 142980
MIM: 142981
MIM: 611576
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HOXD-AS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs55799551] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HOXD-AS2
- Gene Name
- HOXD cluster antisense RNA 2
There are no transcripts associated with this gene.
- Gene
- HOXD3
- Gene Name
- homeobox D3
- Gene
- HOXD4
- Gene Name
- homeobox D4
There are no transcripts associated with this gene.
- Gene
- MIR10B
- Gene Name
- microRNA 10b
There are no transcripts associated with this gene.
View Full Product Details